Learn More
enQuireBio™ Recombinant Human S-arrestin Protein
A DNA sequence encoding the Homo sapiens (Human) S-arrestin, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP9381-ye-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16088211
  • 17300.00 DKK / 500µg
Estimated Shipment: 02-07-2024
to see stock.
Specifications
NP_000532.2
S-arrestin Protein
Research Use Only
SAG
Recombinant Protein
Yeast
MAASGKTSKS EPNHVIFKKI SRDKSVTIYL GNRDYIDHVS QVQPVDGVVL VDPDLVKGKK VYVTLTCAFR YGQEDIDVIG LTFRRDLYFS RVQVYPPVGA ASTPTKLQES LLKKLGSNTY PFLLTFPDYL PCSVMLQPAP QDSGKSCGVD FEVKAFATDS TDAEEDKIPK KSSVRLLIRK VQHAPLEMGP QPRAEAAWQF FMSDKPLHLA VSLNKEIYFH GEPIPVTVTV TNNTEKTVKK IKAFVEQVAN VVLYSSDYYV KPVAMEEAQE KVPPNSTLTK TLTLLPLLAN NRERRGIALD GKIKHEDTNL ASSTIIKEGI DRTVLGILVS YQIKVKLTVS GFLGELTSSE VATEVPFRLM HPQPEDPAKE SYQDANLVFE EFARHNLKDA GEAEEGKRDK NDVDE
Greater than 90% as determined by SDS-PAGE.
6295
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol