missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RISC-loading complex subunit TARBP2 Protein
A DNA sequence encoding the Homo sapiens (Human) RISC-loading complex subunit TARBP2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7601-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_004169.3 | |
RISC-loading complex subunit TARBP2 Protein | |
Research Use Only | |
TARBP2 | |
Recombinant Protein | |
E. coli | |
MSEEEQGSGT TTGCGLPSIE QMLAANPGKT PISLLQEYGT RIGKTPVYDL LKAEGQAHQP NFTFRVTVGD TSCTGQGPSK KAAKHKAAEV ALKHLKGGSM LEPALEDSSS FSPLDSSLPE DIPVFTAAAA ATPVPSVVLT RSPPMELQPP VSPQQSECNP VGALQELVVQ KGWRLPEYTV TQESGPAHRK EFTMTCRVER FIEIGSGTSK KLAKRNAAAK MLLRVHTVPL DARDGNEVEP DDDHFSIGVG SRLDGLRNRG PGCTWDSLRN SVGEKILSLR SCSLGSLGAL GPACCRVLSE LSEEQAFHVS YLDIEELSLS GLCQCLVELS TQPATVCHGS ATTREAARGE AARRALQYLK IMAGSK | |
Greater than 80% as determined by SDS-PAGE. |
6895 | |
50 ÎĽg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human RISC-loading complex subunit TARBP2 Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction