missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RING finger protein 11 Protein
A DNA sequence encoding the Homo sapiens (Human) RING finger protein 11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9370-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055187.1 | |
RING finger protein 11 Protein | |
Research Use Only | |
RNF11 | |
Recombinant Protein | |
Yeast | |
GNCLKSPTSD DISLLHESQS DRASFGEGTE PDQEPPPPYQ EQVPVPVYHP TPSQTRLATQ LTEEEQIRIA QRIGLIQHLP KGVYDPGRDG SEKKIRECVI CMMDFVYGDP IRFLPCMHIY HLDCIDDWLM RSFTCPSCME PVDAALLSSY ETN | |
Greater than 90% as determined by SDS-PAGE. |
26994 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |