missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ribosomal protein S6 kinase beta-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Ribosomal protein S6 kinase beta-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8430-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064571

  • 15780.00 DKK / 1mg
Estimated Shipment: 31-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003943.2
Ribosomal protein S6 kinase beta-2 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
6199
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RPS6KB2
Recombinant Protein
E. coli
MAAVFDLDLE TEEGSEGEGE PELSPADACP LAELRAAGLE PVGHYEEVEL TETSVNVGPE RIGPHCFELL RVLGKGGYGK VFQVRKVQGT NLGKIYAMKV LRKAKIVRNA KDTAHTRAER NILESVKHPF IVELAYAFQT GGKLYLILEC LSGGELFTHL EREGIFLEDT ACFYLAEITL ALGHLHSQGI IYRDLKPENI MLSSQGHIKL
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers