missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ribonucleoprotein PTB-binding 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribonucleoprotein PTB-binding 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8116-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_060681.2 | |
Ribonucleoprotein PTB-binding 2 Protein | |
Research Use Only | |
RAVER2 | |
Recombinant Protein | |
E. coli | |
AAAAGDGGGE GGAGLGSAAG LGPGPGLRGQ GPSAEAHEGA PDPMPAALHP EEVAARLQRM QRELSNRRKI LVKNLPQDSN CQEVHDLLKD YDLKYCYVDR NKRTAFVTLL NGEQAQNAIQ MFHQYSFRGK DLIVQLQPTD ALLCITNVPI SFTSEEFEEL VRAYGNIERC FLVYSEVTGH SKGYGFVEYM KKDFAAKARL ELLGRQLGAS ALFAQWMDVN LLASELIHSK CLCIDKLPSD YRDSEELLQI FSSVHKPVFC QLAQDEGSYV GGFAVVEYST AEQAEEVQQA ADGMTIKGSK VQVSFCAPGA PGRSTLAALI AAQRVMHSNQ KGLLPEPNPV QIMKSLNNPA MLQVLLQPQL CGRAVKPAVL GTPHSLPHLM NPSISPAFLH LNKAHQSSVM GNTSNLFLQN LSHIPLAQQQ LMKFENIHTN NKPGLLGEPP AVVLQTALGI GSVLPLKKEL GHHHGEAHKT SSLIPTQTTI TAGMGMLPFF PNQHIAGQAG PGHSNTQEKQ PATVGMAEGN FSGSQPYLQS FPNLAAGSLL VGHHKQQQSQ PKGTEISSGA ASKNQTSLLG EPPKEIRLSK NPYLNLASVL PSVCLSSPAS KTTLHKTGIA SSILDAISQG SESQHALEKC IAYSPPFGDY AQVSSLRNEK RGSSYLISAP EGGSVECVDQ HSQGTGAYYM ETYLKKKRVY | |
Greater than 90% as determined by SDS-PAGE. |
55225 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Ribonucleoprotein PTB-binding 2 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction