missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Resistin / ADSF / RETN Protein
A DNA sequence encoding the Homo sapiens (Human) Resistin / ADSF / RETN, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6599-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001180303.1 | |
Resistin / ADSF / RETN Protein | |
Research Use Only | |
RETN | |
Recombinant Protein | |
E. coli | |
KTLCSMEEAI NERIQEVAGS LIFRAISSIG LECQSVTSRG DLATCPRGFA VTGCTCGSAC GSWDVRAETT CHCQCAGMDW TGARCCRVQP | |
Greater than 90% as determined by SDS-PAGE. |
56729 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |