missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulator of G-protein signaling 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6603-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002914.1 | |
Regulator of G-protein signaling 2 Protein | |
Research Use Only | |
RGS2 | |
Recombinant Protein | |
E. coli | |
MQSAMFLAVQ HDCRPMDKSA GSGHKSEEKR EKMKRTLLKD WKTRLSYFLQ NSSTPGKPKT GKKSKQQAFI KPSPEEAQLW SEAFDELLAS KYGLAAFRAF LKSEFCEENI EFWLACEDFK KTKSPQKLSS KARKIYTDFI EKEAPKEINI DFQTKTLIAQ NIQEATSGCF TTAQKRVYSL MENNSYPRFL ESEFYQDLCK KPQITTEPHA T | |
Greater than 90% as determined by SDS-PAGE. |
5997 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |