missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Regulator of G-protein signaling 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6603-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054480

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002914.1
Regulator of G-protein signaling 2 Protein
Research Use Only
RGS2
Recombinant Protein
E. coli
MQSAMFLAVQ HDCRPMDKSA GSGHKSEEKR EKMKRTLLKD WKTRLSYFLQ NSSTPGKPKT GKKSKQQAFI KPSPEEAQLW SEAFDELLAS KYGLAAFRAF LKSEFCEENI EFWLACEDFK KTKSPQKLSS KARKIYTDFI EKEAPKEINI DFQTKTLIAQ NIQEATSGCF TTAQKRVYSL MENNSYPRFL ESEFYQDLCK KPQITTEPHA T
Greater than 90% as determined by SDS-PAGE.
5997
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers