missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RARRES2 / Chemerin Protein
A DNA sequence encoding the Homo sapiens (Human) RARRES2 / Chemerin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8013-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002880.1 | |
RARRES2 / Chemerin Protein | |
Research Use Only | |
RARRES2 | |
Recombinant Protein | |
E. coli | |
ELTEAQRRGL QVALEEFHKH PPVQWAFQET SVESAVDTPF PAGIFVRLEF KLQQTSCRKR DWKKPECKVR PNGRKRKCLA CIKLGSEDKV LGRLVHCPIE TQVLREAEEH QETQCLRVQR AGEDPHSFYF PGQFAFS | |
Greater than 80% as determined by SDS-PAGE. |
5919 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |