missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPRR Protein
A DNA sequence encoding the Homo sapiens (Human) PTPRR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7619-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
PTPRR Protein | |
Research Use Only | |
PTPRR | |
Recombinant Protein | |
E. coli | |
MILHRLKERF QLSLRQDKEK NQEIHLSPIT LQPALSEAKT VHSMVQPEQA PKVLNVVVDP QGRGAPEIKA TTATSVCPSP FKMKPIGLQK RRGSNVSLTL DMSSLGNIEP FVSIPTPREK VAMEYLQSAS RILTRSQLRD VVASSHLLQS EFMEIPMNFV DPKEIDIPRH GTKNRYKTIL PNPLSRVCLR PKNVTDSLST YINANYIRGY SGKEKAFIAT QGPMINTVDD FWQMVWQEDS PVIVMITKLK EKNEKCVLYW PEKRGIYGKV EVLVISVNEC DNYTIRNLVL KQGSHTQHVK HYWYTSWPDH KTPDSAQPLL QLMLDVEEDR LASQGRGPVV VHCSAGIGRT GCFIATSIGC QQLKEEGVVD ALSIVCQLRM DRGGMVQTSE QYEFVHHALC LYESRLSAET VQ | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |