missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pseudokinase FAM20A Protein
A DNA sequence encoding the Homo sapiens (Human) Pseudokinase FAM20A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9870-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001230675.1 | |
Pseudokinase FAM20A Protein | |
Research Use Only | |
FAM20A | |
Recombinant Protein | |
Yeast | |
QLRPRERPRG CPCTGRASSL ARDSAAAASD PGTIVHNFSR TEPRTEPAGG SHSGSSSKLQ ALFAHPLYNV PEEPPLLGAE DSLLASQEAL RYYRRKVARW NRRHKMYREQ MNLTSLDPPL QLRLEASWVQ FHLGINRHGL YSRSSPVVSK LLQDMRHFPT ISADYSQDEK ALLGACDCTQ IVKPSGVHLK LVLRFSDFGK AMFKPMRQQR DEETPVDFFY FIDFQRHNAE IAAFHLDRIL DFRRVPPTVG RIVNVTKEIL EVTKNEILQS VFFVSPASNV CFFAKCPYMC KTEYAVCGNP HLLEGSLSAF LPSLNLAPRL SVPNPWIRSY TLAGKEEWEV NPLYCDTVKQ IYPYNNSQRL LNVIDMAIFD FLIGNMDRHH YEMFTKFGDD GFLIHLDNAR GFGRHSHDEI SILSPLSQCC MIKKKTLLHL QLLAQADYRL SDVMRESLLE DQLSPVLTEP HLLALDRRLQ TILRTVEGCI VAHGQQSVIV DGPVEQLAPD SGQANLTS | |
Greater than 90% as determined by SDS-PAGE. |
54757 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |