missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSAP / Prosaposin Protein
A DNA sequence encoding the Homo sapiens (Human) PSAP / Prosaposin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6540-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035930.1 | |
PSAP / Prosaposin Protein | |
Research Use Only | |
PSAP | |
Recombinant Protein | |
E. coli | |
SDVYCEVCEF LVKEVTKLID NNKTEKEILD AFDKMCSKLP KSLSEECQEV VDTYGSSILS ILLEEVSPEL VCSMLHLCSG T | |
Greater than 90% as determined by SDS-PAGE. |
5660 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |