missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein DGCR6L Protein
A DNA sequence encoding the Homo sapiens (Human) Protein DGCR6L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8165-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_150282.2 | |
Protein DGCR6L Protein | |
Research Use Only | |
DGCR6L | |
Recombinant Protein | |
E. coli | |
MERYAAALEE VADGARQQER HYQLLSALQS LVKELPSSFQ QRLSYTTLSD LALALLDGTV FEIVQGLLEI QHLTEKSLYN QRLRLQNEHR VLRQALRQKH QEAQQACRPH NLPVVQAAQQ RELEAVEHRI REEQRAMDQK IILELDRKVA DQQSTLEKAG VAGFYVTTNP QELMLQMNLL ELIRKLQQRG CRAGNAALGL GGPWQSPAAQ CDQKGSPVPP | |
Greater than 90% as determined by SDS-PAGE. |
85359 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |