missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Human Protein delta homolog 2 Protein

Artikelnummer. 16070601
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16070601

Brand: enQuireBio™ QP7755ec1mg

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

A DNA sequence encoding the Homo sapiens (Human) Protein delta homolog 2, was expressed in the hosts and tags indicated.

Tekniske data

Adgangsnummer NP_001273584.1
Gen-id (Entrez) 65989
Navn Protein delta homolog 2 Protein
Mængde 1 mg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol DLK2
Protein længde Extracellular Domain
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter E. coli
Protein tag GST
Sekvens DDCSSHCDLA HGCCAPDGSC RCDPGWEGLH CERCVRMPGC QHGTCHQPWQ CICHSGWAGK FCDKDEHICT TQSPCQNGGQ CMYDGGGEYH CVCLPGFHGR DCERKAGPCE QAGSPCRNGG QCQDDQGFAL NFTCRCLVGF VGARCEVNVD DCLMRPCANG ATCLDGINRF SCLCPEGFAG RFCTINLDDC ASRPCQRGAR CRDRVHDFDC LCPSGYGGKT CELVLPVPDP PTTVDTPLGP TSAVVVPATG PAPHSAGAGL LRISVKEVVR RQEAGLGEPS
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.