missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein ABHD18 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein ABHD18, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7525-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
Protein ABHD18 Protein | |
Research Use Only | |
C4orf29 | |
Recombinant Protein | |
E. coli | |
RPEDLKRLFE FRKMIGNRER CQNLVSSDYP VHIDKIEEQS DCKILDGHFV SPMAHYVPDI MPIESVIARF QFIVPKEWNS KYRPVCIHLA GTGDHHYWRR RTLMARPMIK EARMASLLLE NPYYGCRKPK DQVRSSLKNV SDLFVMGGAL VLESAALLHW LEREGYGPLG MTGISMGGHM ASLAVSNWPK PMPLIPCLSW STASGVFTTT DSFKMGQEFV KHFTSSADKL TNLNLVSRTL NLDISNQVVS QKPADCHNSS KTSVSATSEG LLLQDTSKMK RFNQTLSTNK SGYTSRNPQS YHLLSKEQSR NSLRKESLIF MKGVMDECTH VANFSVPVDP SLIIVVQAKE DAYIPRTGVR SLQEIWPGCE IRYLEGGHIS AYLFKQGLFR | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |