missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proteasome subunit beta type-10 Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-10, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_002792.1 |
---|---|
Gene ID (Entrez) | 5699 |
Name | Proteasome subunit beta type-10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16054050
|
enQuireBio™
QP6544-EC-100UG |
100 μg |
3450.00 DKK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16074050
|
enQuireBio™
QP6544-EC-1MG |
1 mg |
15565.00 DKK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064050
|
enQuireBio™
QP6544-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084050
|
enQuireBio™
QP6544-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094050
|
enQuireBio™
QP6544-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004060
|
enQuireBio™
QP6544-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002792.1 | |
Proteasome subunit beta type-10 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5699 | |
Research Use Only | |
PSMB10 | |
Recombinant Protein | |
E. coli | |
TTIAGLVFQD GVILGADTRA TNDSVVADKS CEKIHFIAPK IYCCGAGVAA DAEMTTRMVA SKMELHALST GREPRVATVT RILRQTLFRY QGHVGASLIV GGVDLTGPQL YGVHPHGSYS RLPFTALGSG QDAALAVLED RFQPNMTLEA AQGLLVEAVT AGILGDLGSG GNVDACVITK TGAKLLRTLS SPTEPVKRSG RYHFVPGTTA VLTQTVKPLT LELVEETVQA MEVE | |
Greater than 80% as determined by SDS-PAGE. |