missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proteasome inhibitor PI31 subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasome inhibitor PI31 subunit, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7980-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001310336.1 | |
Proteasome inhibitor PI31 subunit Protein | |
Research Use Only | |
PSMF1 | |
Recombinant Protein | |
E. coli | |
MAGLEVLFAS AAPAITCRQD ALVCFLHWEV VTHGYFGLGV GDQPGPNDKK SELLPAGWNN NKDLYVLRYE YKDGSRKLLV KAITVESSMI LNVLEYGSQQ VADLTLNLDD YIDAEHLGDF HRTYKNSEEL RSRIVSGIIT PIHEQWEKAN VSSPHREFPP ATAREVDPLR IPPHHPHTSR QPPWCDPLGP FVVGGEDLDP FGPRRGGMIV DPLRSGFPRA LIDPSSGLPN RLPPGAVPPG ARFDPFGPIG TSPPGPNPDH LPPPGYDDMY L | |
Greater than 90% as determined by SDS-PAGE. |
9491 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |