missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proteasome inhibitor PI31 subunit Protein

A DNA sequence encoding the Homo sapiens (Human) Proteasome inhibitor PI31 subunit, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7980-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16071951

  • 15565.00 DKK / 1mg
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001310336.1
Proteasome inhibitor PI31 subunit Protein
Research Use Only
PSMF1
Recombinant Protein
E. coli
MAGLEVLFAS AAPAITCRQD ALVCFLHWEV VTHGYFGLGV GDQPGPNDKK SELLPAGWNN NKDLYVLRYE YKDGSRKLLV KAITVESSMI LNVLEYGSQQ VADLTLNLDD YIDAEHLGDF HRTYKNSEEL RSRIVSGIIT PIHEQWEKAN VSSPHREFPP ATAREVDPLR IPPHHPHTSR QPPWCDPLGP FVVGGEDLDP FGPRRGGMIV DPLRSGFPRA LIDPSSGLPN RLPPGAVPPG ARFDPFGPIG TSPPGPNPDH LPPPGYDDMY L
Greater than 90% as determined by SDS-PAGE.
9491
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers