missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proteasomal ubiquitin receptor ADRM1 Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasomal ubiquitin receptor ADRM1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7545-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001268366.1 | |
Proteasomal ubiquitin receptor ADRM1 Protein | |
Research Use Only | |
ADRM1 | |
Recombinant Protein | |
E. coli | |
TTSGALFPSL VPGSRGASNK YLVEFRAGKM SLKGTTVTPD KRKGLVYIQQ TDDSLIHFCW KDRTSGNVED DLIIFPDDCE FKRVPQCPSG RVYVLKFKAG SKRLFFWMQE PKTDQDEEHC RKVNEYLNNP PMPGALGASG SSGHELSALG GEGGLQSLLG NMSHSQLMQL IGPAGLGGLG GLGALTGPGL ASLLGSSGPP GSSSSSSSRS QSAAVTPSST TSSTRATPAP SAPAAASATS PSPAPSSGNG ASTAASPTQP IQLSDLQSIL ATMNVPAGPA GGQQVDLASV LTPEIMAPIL ANADVQERLL PYLPSGESLP QTADEIQNTL TSPQFQQALG MFSAALASGQ LGPLMCQFGL PAEAVEAANK GDVEAFAKAM QNNAKPEQKE GDTKDKKDEE EDMSLD | |
Greater than 90% as determined by SDS-PAGE. |
11047 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |