missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Probable G-protein coupled receptor 75 Protein

A DNA sequence encoding the Homo sapiens (Human) Probable G-protein coupled receptor 75, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6113-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070890

  • 4195.00 DKK / 100µg
Estimated Shipment: 24-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006785.1
Probable G-protein coupled receptor 75 Protein
100 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
10936
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GPR75
Recombinant Protein
Yeast
NPFIYSRNSA GLRRKVLWCL QYIGLGFFCC KQKTRLRAMG KGNLEVNRNK SSHHETNSAY MLSPKPQKKF VDQACGPSHS KESMVSPKIS AGHQHCGQSS STPINTRIEP YYSIYNSSPS QEESSPCNLQ PVNSFGFANS YIAMHYHTTN DLVQEYDSTS AKQIPVPSV
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers