missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PNPLA2 Protein
A DNA sequence encoding the Homo sapiens (Human) PNPLA2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7868-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_065109.1 | |
PNPLA2 Protein | |
Research Use Only | |
PNPLA2 | |
Recombinant Protein | |
E. coli | |
MFPREKTWNI SFAGCGFLGV YYVGVASCLR EHAPFLVANA THIYGASAGA LTATALVTGV CLGEAGAKFI EVSKEARKRF LGPLHPSFNL VKIIRSFLLK VLPADSHEHA SGRLGISLTR VSDGENVIIS HFNSKDELIQ ANVCSGFIPV YCGLIPPSLQ GVRYVDGGIS DNLPLYELKN TITVSPFSGE SDICPQDSST NIHELRVTNT SIQFNLRNLY RLSKALFPPE PLVLREMCKQ GYRDGLRFLQ RNGLLNRPNP LLALPPARPH GPEDKDQAVE SAQAEDYSQL PGEDHILEHL PARLNEALLE ACVEPTDLLT TLSNMLPVRL ATAMMVPYTL PLESALSFTI RLLEWLPDVP EDIRWMKEQT GSICQYLVMR AKRKLGRHLP SRLPEQVELR RVQSLPSVPL SCAAYREALP GWMRNNLSLG DALAKWEECQ RQLLLGLFCT NVAFPPEALR MRAPADPAPA PADPASPQHQ LAGPAPLLST PAPEARPVIG ALGL | |
Greater than 80% as determined by SDS-PAGE. |
57104 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |