missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Perforin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Perforin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6525-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001076585.1 | |
Perforin-1 Protein | |
Research Use Only | |
Prf1 | |
Recombinant Protein | |
Yeast | |
PCHTAARSEC KRSHKFVPGA WLAGEGVDVT SLRRSGSFPV DTQRFLRPDG TCTLCENALQ EGTLQRLPLA LTNWRAQGSG CQRHVTRAKV SSTEAVARDA ARSIRNDWKV GLDVTPKPTS NVHVSVAGSH SQAANFAAQK THQDQYSFST DTVECRFYSF HVVHTPPLHP DFKRALGDLP HHFNASTQPA YLRLISNYGT HFIRAVELGG RISALTALRT CELALEGLTD NEVEDCLTVE AQVNIGIHGS ISAEAKACEE KKKKHKMTAS FHQTYRERHS EVVGGHHTSI NDLLFGIQAG PEQYSAWVNS LPGSPGLVDY TLEPLHVLLD SQDPRREALR RALSQYLTDR ARWRDCSRPC PPGRQKSPRD PCQCVCHGSA VTTQDCCPRQ RGLAQLEVTF IQAWGLWGDW FTATDAYVKL FFGGQELRTS TVWDNNNPIW SVRLDFGDVL LATGGPLRLQ VWDQDSGRDD DLLGTCDQAP KSGSHEVRCN LNHGHLKFRY HARCLPHLGG GTCLDYVPQM LLGEPPGNRS GAVW | |
Greater than 80% as determined by SDS-PAGE. |
5551 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |