missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Perforin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Perforin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6525-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003930

  • 17300.00 DKK / 500µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001076585.1
Perforin-1 Protein
Research Use Only
Prf1
Recombinant Protein
Yeast
PCHTAARSEC KRSHKFVPGA WLAGEGVDVT SLRRSGSFPV DTQRFLRPDG TCTLCENALQ EGTLQRLPLA LTNWRAQGSG CQRHVTRAKV SSTEAVARDA ARSIRNDWKV GLDVTPKPTS NVHVSVAGSH SQAANFAAQK THQDQYSFST DTVECRFYSF HVVHTPPLHP DFKRALGDLP HHFNASTQPA YLRLISNYGT HFIRAVELGG RISALTALRT CELALEGLTD NEVEDCLTVE AQVNIGIHGS ISAEAKACEE KKKKHKMTAS FHQTYRERHS EVVGGHHTSI NDLLFGIQAG PEQYSAWVNS LPGSPGLVDY TLEPLHVLLD SQDPRREALR RALSQYLTDR ARWRDCSRPC PPGRQKSPRD PCQCVCHGSA VTTQDCCPRQ RGLAQLEVTF IQAWGLWGDW FTATDAYVKL FFGGQELRTS TVWDNNNPIW SVRLDFGDVL LATGGPLRLQ VWDQDSGRDD DLLGTCDQAP KSGSHEVRCN LNHGHLKFRY HARCLPHLGG GTCLDYVPQM LLGEPPGNRS GAVW
Greater than 80% as determined by SDS-PAGE.
5551
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers