missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDZ and LIM domain protein 1
A DNA sequence encoding the Homo sapiens (Human) PDZ and LIM domain protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7666-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001107579.1 | |
PDZ and LIM domain protein 1 | |
Research Use Only | |
PDLIM3 | |
Recombinant Protein | |
E. coli | |
MPQTVILPGP APWGFRLSGG IDFNQPLVIT RITPGSKAAA ANLCPGDVIL AIDGFGTESM THADAQDRIK AAAHQLCLKI DRGETHLWSP QVSEDGKAHP FKINLESEPQ EFKPIGTAHN RRAQPFVAAA NIDDKRQVVS ASYNSPIGLY STSNIQDALH GQLRGLIPSS PQKTGTTLNT SIIFGPNLS | |
Greater than 90% as determined by SDS-PAGE. |
27295 | |
500 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human PDZ and LIM domain protein 1 > 500μg
Spot an opportunity for improvement?Share a Content Correction