missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Osteonectin / SPARC Protein

A DNA sequence encoding the Homo sapiens (Human) Osteonectin / SPARC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8664-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065841

  • 15780.00 DKK / 1mg
Estimated Shipment: 31-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001296372.1
Osteonectin / SPARC Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6678
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SPARC
Recombinant Protein
E. coli
APQQEALPDE TEVVEETVAE VTEVSVGANP VQVEVGEFDD GAEETEEEVV AENPCQNHHC KHGKVCELDE NNTPMCVCQD PTSCPAPIGE FEKVCSNDNK TFDSSCHFFA TKCTLEGTKK GHKLHLDYIG PCKYIPPCLD SELTEFPLRM RDWLKNVLVT LYERDEDNNL LTEKQKLRVK KIHENEKRLE AGDHPVELLA RDFEKNYNMY IFPVHWQFGQ LDQHPIDGYL SHTELAPLRA PLIPMEHCTT RFFETCDLDN DKYIALDEWA GCFGIKQKDI DKDLVI
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers