missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Oncostatin M / OSM Protein
A DNA sequence encoding the Homo sapiens (Human) Oncostatin M / OSM, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8670-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001306037.1 | |
Oncostatin M / OSM Protein | |
Research Use Only | |
OSM | |
Recombinant Protein | |
E. coli | |
AAIGSCSKEY RVLLGQLQKQ TDLMQDTSRL LDPYIRIQGL DVPKLREHCR ERPGAFPSEE TLRGLGRRGF LQTLNATLGC VLHRLADLEQ RLPKAQDLER SGLNIEDLEK LQMARPNILG LRNNIYCMAQ LLDNSDTAEP TKAGRGASQP PTPTPASDAF QRKLEGCRFL HGYHRFMHSV GRVFSKWGES PNRSR | |
Greater than 90% as determined by SDS-PAGE. |
5008 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Oncostatin M / OSM Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction