missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleolar transcription factor 1 Protein
A DNA sequence encoding the Homo sapiens(Human) Nucleolar transcription factor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9990-iv-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001070151.1 | |
Nucleolar transcription factor 1 Protein | |
Research Use Only | |
UBTF | |
Recombinant Protein | |
E. coli | |
MNGEADCPTD LEMAAPKGQD RWSQEDMLTL LECMKNNLPS NDSSKFKTTE SHMDWEKVAF KDFSGDMCKL KWVEISNEVR KFRTLTELIL DAQEHVKNPY KGKKLKKHPD FPKKPLTPYF RFFMEKRAKY AKLHPEMSNL DLTKILSKKY KELPEKKKMK YIQDFQREKQ EFERNLARFR EDHPDLIQNA KKSDIPEKPK TPQQLWYTHE KKVYLKVRPD ATTKEVKDSL GKQWSQLSDK KRLKWIHKAL EQRKEYEEIM RDYIQKHPEL NISEEGITKS TLTKAERQLK DKFDGRPTKP PPNSYSLYCA ELMANMKDVP STERMVLCSQ QWKLLSQKEK DAYHKKCDQK KKDYEVELLR FLESLPEEEQ QRVLGEEKML NINKKQATSP ASKKPAQEGG KGGSEKPKRP VSAMFIFSEE KRRQLQEERP ELSESELTRL LARMWNDLSE KKKAKYKARE AALKAQSERK PGGEREERGK LPESPKRAEE IWQQSVIGDY LARFKNDRVK ALKAMEMTWN NMEKKEKLMW IKKAAEDQKR YERELSEMRA PPAATNSSKK MKFQGEPKKP PMNGYQKFSQ ELLSNGELNH LPLKERMVEI GSRWQRISQS QKEHYKKLAE EQQKQYKVHL DLWVKSLSPQ DRAAYKEYIS NKRKSMTKLR GPNPKSSRTT LQSKSESEED DEEDEDDEDE DEEEEDDENG DSSEDGGDSS ESSSEDESED GDENEEDDED EDDDEDDDED EDNESEGSSS SSSSSGDSSD SDSN | |
Greater than 85% as determined by SDS-PAGE. |
7343 | |
50 μg | |
Not determined. | |
Full length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |