missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear cap-binding protein subunit 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear cap-binding protein subunit 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6407-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001036005.1 | |
Nuclear cap-binding protein subunit 2 Protein | |
Research Use Only | |
NCBP2 | |
Recombinant Protein | |
E. coli | |
SGGLLKALRS DSYVELSQYR DQHFRGDNEE QEKLLKKSCT LYVGNLSFYT TEEQIYELFS KSGDIKKIIM GLDKMKKTAC GFCFVEYYSR ADAENAMRYI NGTRLDDRII RTDWDAGFKE GRQYGRGRSG GQVRDEYRQD YDAGRGGYGK LAQNQ | |
Greater than 80% as determined by SDS-PAGE. |
22916 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |