missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Negative elongation factor B Protein
A DNA sequence encoding the Homo sapiens (Human) Negative elongation factor B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8455-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_056271.3 | |
Negative elongation factor B Protein | |
Research Use Only | |
COBRA1 | |
Recombinant Protein | |
E. coli | |
LGVANGEDLK ETLTNCTEPL KAIEQFQTEN GVLLPSLQSA LPFLDLHGTP RLEFHQSVFD ELRDKLLERV SAIASEGKAE ERYKKLEDLL EKSFSLVKMP SLQPVVMCVM KHLPKVPEKK LKLVMADKEL YRACAVEVKR QIWQDNQALF GDEVSPLLKQ YILEKESALF STELSVLHNF FSPSPKTRRQ GE | |
Greater than 90% as determined by SDS-PAGE. |
25920 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Negative elongation factor B Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction