missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myosin-binding protein C, cardiac-type Protein

A DNA sequence encoding the Homo sapiens (Human) Myosin-binding protein C, cardiac-type, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7532-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16079210

  • 3455.00 DKK / 100µg
Estimated Shipment: 24-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000247.2
Myosin-binding protein C, cardiac-type Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
4607
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Mybpc3
Recombinant Protein
E. coli
MPEPGKKPVS AFSKKPRSVE VAAGSPAVFE AETERAGVKV RWQRGGSDIS ASNKYGLATE GTRHTLTVRE VGPADQGSYA VIAGSSKVKF DLKVIEAEKA EPMLAPAPAP AEATGAPGEA PAPAAELGES APSPKGSSSA ALNGPTPGAP DDPIGLFVMR PQDGEVTVGG SITFSARVAG ASLLKPPVVK WFKGKWVDLS SKVGQHLQLH DSYDRASKVY LFELHITDAQ PAFTGSYRCE VSTKDKFDCS NFNLTVHEAM GTGDLDLLSA FRRTSLAGGG RRISDSHEDT GILDFSSLLK KRDSFRTPRD SKLEAPAEED VWEILRQA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers