missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MSRA Protein
A DNA sequence encoding the Homo sapiens (Human) MSRA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8191-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129142.1 | |
MSRA Protein | |
Research Use Only | |
MSRA | |
Recombinant Protein | |
E. coli | |
GNSASNIVSP QEALPGRKEQ TPVAAKHHVN GNRTVEPFPE GTQMAVFGMG CFWGAERKFW VLKGVYSTQV GFAGGYTSNP TYKEVCSEKT GHAEVVRVVY QPEHMSFEEL LKVFWENHDP TQGMRQGNDH GTQYRSAIYP TSAKQMEAAL SSKENYQKVL SEHGFGPITT DIREGQTFYY AEDYHQQYLS KNPNGYCGLG GTGVSCPVGI KK | |
Greater than 90% as determined by SDS-PAGE. |
4482 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |