missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MSRA Protein

A DNA sequence encoding the Homo sapiens (Human) MSRA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8191-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16053171

  • 15780.00 DKK / 1mg
Estimated Shipment: 03-08-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001129142.1
MSRA Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
4482
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MSRA
Recombinant Protein
E. coli
GNSASNIVSP QEALPGRKEQ TPVAAKHHVN GNRTVEPFPE GTQMAVFGMG CFWGAERKFW VLKGVYSTQV GFAGGYTSNP TYKEVCSEKT GHAEVVRVVY QPEHMSFEEL LKVFWENHDP TQGMRQGNDH GTQYRSAIYP TSAKQMEAAL SSKENYQKVL SEHGFGPITT DIREGQTFYY AEDYHQQYLS KNPNGYCGLG GTGVSCPVGI KK
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers