missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human mRNA-decapping enzyme 1B Protein
A DNA sequence encoding the Homo sapiens(Human) mRNA-decapping enzyme 1B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9835-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_689853.3 | |
mRNA-decapping enzyme 1B Protein | |
Research Use Only | |
DCP1B | |
Recombinant Protein | |
Yeast | |
AAVAAGGLVG KGRDISLAAL QRHDPYINRI VDVASQVALY TFGHRANEWE KTDVEGTLFV YTRSASPKHG FTIMNRLSME NRTEPITKDL DFQLQDPFLL YRNARLSIYG IWFYDKEECQ RIAELMKNLT QYEQLKAHQG TGAGISPVIL NSGEGKEVDI LRMLIKAKDE YTKCKTCSEP KKITSSSAIY DNPNLIKPIP VKPSENQQQR IPQPNQTLDP EPQHLSLTAL FGKQDKATCQ ETVEPPQTLH QQQQQQQQQQ EKLPIRQGVV RSLSYEEPRR HSPPIEKQLC PAIQKLMVRS ADLHPLSELP ENRPCENGST HSAGEFFTGP VQPGSPHNIG TSRGVQNASR TQNLFEKLQS TPGAANKCDP STPAPASSAA LNRSRAPTSV TPVAPGKGLA QPPQAYFNGS LPPQTVGHQA HGREQSTLPR QTLPISGSQT GSSGVISPQE LLKKLQIVQQ EQQLHASNRP ALAAKFPVLA QSSGTGKPLE SWINKTPNTE QQTPLFQVIS PQRIPATAAP SLLMSPMVFA QPTSVPPKER ESGLLPVGGQ EPPAAATSLL LPIQSPEPSV ITSSPLTKLQ LQEALLYLIQ NDDNFLNIIY EAYLFSMTQA AMKKTM | |
Greater than 85% as determined by SDS-PAGE. |
196513 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |