Få mere at vide
Novus Biologicals™ Recombinant Human MMP-9 Protein
Artikelnummer. 18161930 Shop alle Bio Techne produkter
Skift visning
Mængde:
50 μg
Artikelnummer. Mængde
18161930 50 μg
1 options
Artikelnummer. 18161930 Leverandør Novus Biologicals™ Leverandørnr. NBP19960450UG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified. Generating reliable and reproducible results.
Recombinant protein for Human MMP-9 corresponding to the catalytic domain The Recombinant Human MMP-9 Protein is derived from E. coli. The Recombinant Human MMP-9 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Tekniske data
Koncentration Lyoph
Til brug med (applikation) SDS-PAGE
Formulering Lyophilized with no additives.
Gen-id (Entrez) 4318
Molekylvægt (g/mol) 39.0 kDa
Mængde 50 μg
Kilde E. coli
Opbevaringskrav Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
Gen symbol MMP9
Rekonstitution Reconstitute in sterile PBS to 0.1 - 1.0 mg/mL. This solution can then be diluted into other aqueous buffers andstored at 4°C for 1 week or -20°C for long-term storage.
Buffer Lyophilized with no additives.
Endotoksin niveau <0.1 ng/μg
Renhed eller kvalitetsklasse >95%, by SDS-PAGE