missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Microfibrillar-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Microfibrillar-associated protein 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7607-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001284638.1 | |
Microfibrillar-associated protein 1 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
8076 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
MFAP5 | |
Recombinant Protein | |
E. coli | |
IPLGVNSQRG DDVTQATPET FTEDPNLVND PATDETVLAV LADIAPSTDD LASLSEKNTT AECWDEKFTC TRLYSVHRPV KQCIHQLCFT SLRRMYIVNK EICSRLVCKE HEAMKDELCR QMAGLPPRRL RRSNYFRLPP CENVDLQRPN GL |