missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Human mGluR4 Protein
Klik for at se tilgængelige muligheder
Mængde:
100 μg
Beskrivelse
The Recombinant Human mGluR4 Protein is derived from CHO.
Tekniske data
Tekniske data
| Adgangsnummer | Q14833-1 |
| Karakterisering | Bioactivity not tested |
| Til brug med (applikation) | Flow Cytometry |
| Formulering | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gen-id (Entrez) | 2914 |
| Molekylvægt (g/mol) | 55kDa |
| Mængde | 100 μg |
| Kilde | Chinese Hamster Ovary cell line,CHO-derived human mGluR4 protein Lys33-Ser518,with a C-terminal 6-His tag |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoksin koncentration | <0.10EU per 1μg of the protein by the LAL method. |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion