missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Methylosome protein 50 Protein
A DNA sequence encoding the Homo sapiens (Human) Methylosome protein 50, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8020-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001303991.1 | |
Methylosome protein 50 Protein | |
Research Use Only | |
WDR77 | |
Recombinant Protein | |
E. coli | |
MRKETPPPLV PPAAREWNLP PNAPACMERQ LEAARYRSDG ALLLGASSLS GRCWAGSLWL FKDPCAAPNE GFCSAGVQTE AGVADLTWVG ERGILVASDS GAVELWELDE NETLIVSKFC KYEHDDIVST VSVLSSGTQA VSGSKDICIK VWDLAQQVVL SSYRAHAAQV TCVAASPHKD SVFLSCSEDN RILLWDTRCP KPASQIGCSA PGYLPTSLAW HPQQSEVFVF GDENGTVSLV DTKSTSCVLS SAVHSQCVTG LVFSPHSVPF LASLSEDCSL AVLDSSLSEL FRSQAHRDFV RDATWSPLNH SL | |
Greater than 90% as determined by SDS-PAGE. |
79084 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |