missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Metallothionein-1F Protein

A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1F, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9291-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16078021

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001288201.1
Metallothionein-1F Protein
Research Use Only
MT1F
Recombinant Protein
Yeast
MDPNCSCAAG VSCTCAGSCK CKECKCTSCK KSCCSCCPVG CSKCAQGCVC KGASEKCSC
Greater than 90% as determined by SDS-PAGE.
4494
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers