missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MART-1 / MLANA Protein

A DNA sequence encoding the Homo sapiens (Human) MART-1 / MLANA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7586-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16089570

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_005502.1
MART-1 / MLANA Protein
Research Use Only
MLANA
Recombinant Protein
E. coli
MPREDAHFIY GYPKKGHGHS YTTAEEAAGI GILTVILGVL LLIGCWYCRR RNGYRALMDK SLHVGTQCAL TRRCPQEGFD HRDSKVSLQE KNCEPVVPNA PPAYEKLSAE QSPPPYSP
Greater than 90% as determined by SDS-PAGE.
2315
500 μg
Not determined.
Full Length
Human
His-B2M
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers