missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAP3K7 C-terminal-like Protein
A DNA sequence encoding the Homo sapiens (Human) MAP3K7 C-terminal-like protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7247-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001273546.1 | |
MAP3K7 C-terminal-like Protein | |
Research Use Only | |
MAP3K7CL | |
Recombinant Protein | |
E. coli | |
MISTARVPAD KPVRIAFSLN DASDDTPPED SIPLVFPELD QQLQPLPPCH DSEESMEVFK QHCQIAEEYH EVKKEITLLE QRKKELIAKL DQAEKEKVDA AELVREFEAL TEENRTLRLA QSQCVEQLEK LRIQYQKRQG SS | |
Greater than 90% as determined by SDS-PAGE. |
56911 | |
50 μg | |
Not determined. | |
Full Length of Isoform D | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human MAP3K7 C-terminal-like Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction