missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Malate dehydrogenase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Malate dehydrogenase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6352-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001269332.1 | |
Malate dehydrogenase, mitochondrial Protein | |
Research Use Only | |
MDH2 | |
Recombinant Protein | |
E. coli | |
AKVAVLGASG GIGQPLSLLL KNSPLVSRLT LYDIAHTPGV AADLSHIETK AAVKGYLGPE QLPDCLKGCD VVVIPAGVPR KPGMTRDDLF NTNATIVATL TAACAQHCPE AMICVIANPV NSTIPITAEV FKKHGVYNPN KIFGVTTLDI VRANTFVAEL KGLDPARVNV PVIGGHAGKT IIPLISQCTP KVDFPQDQLT ALTGRIQEAG TEVVKAKAGA GSATLSMAYA GARFVFSLVD AMNGKEGVVE CSFVKSQETE CTYFSTPLLL GKKGIEKNLG IGKVSSFEEK MISDAIPELK ASIKKGEDFV KTLK | |
Greater than 90% as determined by SDS-PAGE. |
4191 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |