missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ly6 / PLAUR domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Ly6/PLAUR domain-containing protein 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7780-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16080751

  • 4195.00 DKK / 100µg
Estimated Shipment: 31-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001182614.1
Ly6 / PLAUR domain-containing protein 1
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
130574
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LYPD6
Recombinant Protein
Yeast
AQSRDFTVKD IIYLHPSTTP YPGGFKCFTC EKAADNYECN RWAPDIYCPR ETRYCYTQHT MEVTGNSISV TKRCVPLEEC LSTGCRDSEH EGHKVCTSCC EGNICNLPLP RNETDATFAT TSPINQTNGH PRCMSVIVSC LWLWLGLML
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers