missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ly6 / PLAUR domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Ly6/PLAUR domain-containing protein 6, was expressed in the hosts and tags indicated.

1500.00 DKK - 15995.00 DKK

Specifications

Accession Number NP_001182614.1
Gene ID (Entrez) 130574
Name Ly6 / PLAUR domain-containing protein 1
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16090751
View Documents
enQuireBio™
QP7780-ye-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16030761
View Documents
enQuireBio™
QP7780-ye-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16080751
View Documents
enQuireBio™
QP7780-ye-100ug
100 ÎĽg
4645.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16010761
View Documents
enQuireBio™
QP7780-ye-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16020761
View Documents
enQuireBio™
QP7780-ye-500ug
500 ÎĽg
12590.00 DKK
500µg
Please to purchase this item. Need a web account? Register with us today!
16000761
View Documents
enQuireBio™
QP7780-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_001182614.1
Ly6 / PLAUR domain-containing protein 1
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
130574
Research Use Only
LYPD6
Recombinant Protein
Yeast
AQSRDFTVKD IIYLHPSTTP YPGGFKCFTC EKAADNYECN RWAPDIYCPR ETRYCYTQHT MEVTGNSISV TKRCVPLEEC LSTGCRDSEH EGHKVCTSCC EGNICNLPLP RNETDATFAT TSPINQTNGH PRCMSVIVSC LWLWLGLML
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Ly6 / PLAUR domain-containing protein 1

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.