missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lck Kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Lck Kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6288-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001036236.1 | |
Lck Kinase Protein | |
Research Use Only | |
LCK | |
Recombinant Protein | |
E. coli | |
MGCGCSSHPE DDWMENIDVC ENCHYPIVPL DGKGTLLIRN GSEVRDPLVT YEGSNPPASP LQDNLVIALH SYEPSHDGDL GFEKGEQLRI LEQSGEWWKA QSLTTGQEGF IPFNFVAKAN SLEPEPWFFK NLSRKDAERQ LLAPGNTHGS FLIRESESTA GSFSLSVRDF DQNQGEVVKH YKIRNLDNGG FYISPRITFP GLHELVRHYT NASDGLCTRL SRPCQTQKPQ KPWWEDEWEV PRETLKLVER LGAGQFGEVW MGYYNGHTKV AVKSLKQGSM SPDAFLAEAN LMKQLQHQRL VRLYAVVTQE PIYIITEYME NDTLLDSQLE EKGLGASPWG NLGQQLLLLP TGSLVDFLKT PSGIKLTINK LLDMAAQIAE GMAFIEERNY IHRDLRAANI LVSDTLSCKI ADFGLARLIE DNEYTAREGA KFPIKWTAPE AINYGTFTIK SDVWSFGILL TEIVTHGRIP YPGMTNPEVI QNLERGYRMV RPDNCPEELY QLMRLCWKER PEDRPTFDYL RSVLEDFFTA TEGQYQPQP | |
Greater than 90% as determined by SDS-PAGE. |
3932 | |
200 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |