missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LCAT Protein
A DNA sequence encoding the Homo sapiens (Human) LCAT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8712-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000220.1 | |
LCAT Protein | |
Research Use Only | |
LCAT | |
Recombinant Protein | |
E. coli | |
FWLLNVLFPP HTTPKAELSN HTRPVILVPG CLGNQLEAKL DKPDVVNWMC YRKTEDFFTI WLDLNMFLPL GVDCWIDNTR VVYNRSSGLV SNAPGVQIRV PGFGKTYSVE YLDSSKLAGY LHTLVQNLVN NGYVRDETVR AAPYDWRLEP GQQEEYYRKL AGLVEEMHAA YGKPVFLIGH SLGCLHLLYF LLRQPQAWKD RFIDGFISLG APWGGSIKPM LVLASGDNQG IPIMSSIKLK EEQRITTTSP WMFPSRMAWP EDHVFISTPS FNYTGRDFQR FFADLHFEEG WYMWLQSRDL LAGLPAPGVE VYCLYGVGLP TPRTYIYDHG FPYTDPVGVL YEDGDDTVAT RSTELCGLWQ GRQPQPVHLL PLHGIQHLNM VFSNLTLEHI NAILLGAYRQ GPPASPTAS | |
Greater than 90% as determined by SDS-PAGE. |
3931 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |