missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLK5 Protein
A cDNA sequence encoding the KLK5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12507-HIS-INSECT-2ug
Additional Details : Weight : 0.01000kg
Specifications
25818 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MGSHMIINGS DCDMHTQPWQ AALLLRPNQL YCGAVLVHPQ WLLTAAHCRK KVFRVRLGHY SLSPVYESGQ QMFQGVKSIP HPGYSHPGHS NDLMLIKLNR RIRPTKDVRP INVSSHCPSA GTKCLVSGWG TTKSPQVHFP KVLQCLNISV LSQKRCEDAY PRQIDDTMFC AGDKAGRDSC QGDSGGPVVC NGSLQGLVSW GDYPCARPNR PGVYTNLCKF TKWIQETIQA NS | |
Greater than 90.0% as determined by SDS-PAGE. |
KLK5 Protein | |
Research Use Only | |
KLK5 | |
Human | |
His | |
The KLK5 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea and 20% glycerol. |