missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Human JAM-2 / JAM-B Protein

Artikelnummer. 16071900
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16071900

Brand: enQuireBio™ QP6247ec1mg

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) JAM-2 / JAM-B, was expressed in the hosts and tags indicated.

Specifications

Adgangsnummer NP_001257336.1
Gen-id (Entrez) 58494
Navn JAM-2 / JAM-B Protein
Mængde 1 mg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol JAM2
Protein længde Extracellular Domain
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter E. coli
Protein tag His
Sekvens FSAPKDQQVV TAVEYQEAIL ACKTPKKTVS SRLEWKKLGR SVSFVYYQQT LQGDFKNRAE MIDFNIRIKN VTRSDAGKYR CEVSAPSEQG QNLEEDTVTL EVLVAPAVPS CEVPSSALSG TVVELRCQDK EGNPAPEYTW FKDGIRLLEN PRLGSQSTNS SYTMNTKTGT LQFNTVSKLD TGEYSCEARN SVGYRRCPGK RMQVDDLNIS
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.