missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Integrin-linked protein kinase Protein

A DNA sequence encoding the Homo sapiens (Human) Integrin-linked protein kinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8343-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16074121

  • 3455.00 DKK / 100µg
Estimated Shipment: 24-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001014794.1
Integrin-linked protein kinase Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
3611
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ILK1
Recombinant Protein
E. coli
MDDIFTQCRE GNAVAVRLWL DNTENDLNQG DDHGFSPLHW ACREGRSAVV EMLIMRGARI NVMNRGDDTP LHLAASHGHR DIVQKLLQYK ADINAVNEHG NVPLHYACFW GQDQVAEDLV ANGALVSICN KYGEMPVDKA KAPLRELLRE RAEKMGQNLN RIPYKDTFWK GTTRTRPRNG TLNKHSGIDF KQLNFLTKLN ENHSGELWKG RWQGNDIVVK VLKVRDWS
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers