missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inositol-trisphosphate 3-kinase B Protein
A DNA sequence encoding the Homo sapiens (Human) Inositol-trisphosphate 3-kinase B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6243-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_002212.3 | |
Inositol-trisphosphate 3-kinase B Protein | |
Research Use Only | |
ITPKB | |
Recombinant Protein | |
E. coli | |
RVEGGSPTLG LLGGSPSAQP GTGNVEAGIP SGRMLEPLPC WDAAKDLKEP QCPPGDRVGV QPGNSRVWQG TMEKAGLAWT RGTGVQSEGT WESQRQDSDA LPSPELLPQD PDKPFLRKAC SPSNIPAVII TDMGTQEDGA LEETQGSPRG NLPLRKLSSS SASSTGFSSS YEDSEEDISS DPERTLDPNS AFLHTLDQQK PRVSKSWRKI KNMVHWSPFV MSFKKKYPWI QLAGHAGSFK AAANGRILKK HCESEQRCLD RLMVDVLRPF VPAYHGDVVK DGERYNQMDD LLADFDSPCV MDCKMGIRTY LEEELTKARK KPSLRKDMYQ KMIEVDPEAP TEEEKAQRAV TKPRYMQWRE TISSTATLGF RIEGIKKEDG TVNRDFKKTK TREQVTEAFR EFTKGNHNIL IAYRDRLKAI RTTLEVSPFF KCHEVIGSSL LFIHDKKEQA KVWMIDFGKT TPLPEGQTLQ HDVPWQEGNR EDGYLSGLNN LVDILTEMSQ DAPLA | |
Greater than 90% as determined by SDS-PAGE. |
3707 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Inositol-trisphosphate 3-kinase B Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction