missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL7 / Interleukin 7 Protein
A DNA sequence encoding the Homo sapiens(Human) IL7 / Interleukin 7 Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9979-iv-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000871.1 | |
IL7 / Interleukin 7 Protein | |
Research Use Only | |
IL7 | |
Recombinant Protein | |
E. coli | |
DCDIEGKDGK QYESVLMVSI DQLLDSMKEI GSNCLNNEFN FFKRHICDAN KEGMFLFRAA RKLRQFLKMN STGDFDLHLL KVSEGTTILL NCTGQVKGRK PAALGEAQPT KSLEENKSLK EQKKLNDLCF LKRLLQEIKT CWNKILMGTK EH | |
Greater than 85% as determined by SDS-PAGE. |
3574 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |